Order Entry
Canada
Orders LinkContactUsLinkComponent
Specimen Mount Holders, Electron Microscopy Sciences

Specimen Mount Holders, Electron Microscopy Sciences

Mount Holder Box for JEOL 840, 10/pk, Box with insert and lid; holds 4 of 12.5mmx12.5mm mounts
  100501-198
Supplier #:  76732
Specimen Mount Holders, Electron Microscopy Sciences
  100501-198
Specimen Mount Holders, Electron Microscopy Sciences
Specimen Mount Holders, Electron Microscopy Sciences

Mount Holder Box for JEOL 840, 10/pk, Box with insert and lid; holds 4 of 12.5mmx12.5mm mounts

 
$118.00
   
Each Pack of 10
 
   
 

Boxes in five configurations are available for storing SEM pin stub mounts and cylinder mounts.

Mount Holder Box, Pin Type with insert and lid holds four of 12.5 mm (1/2") diameter surface and 3.2 mm (1/8") diameter pin mounts.
Mount Holder Box for JEOL with insert and lid holds four of 10 x 10 mm mounts.
Mount Holder Box JEOL 840 with insert and lid holds four of 12.5 x 12.5 mm mounts.
Universal Reversible Mount Holders Box with insert and lid holds 12 of 10 and 15 mm diameter mounts or 12" pin mounts. Overall L x W x H measurements: 12 x 9.5 x 3 cm (43/4 x 35/8 x 1/4").

The box accommodates up to nine mounting stubs of either 25 mm (1") in diameter or a 1/8" diameter pin.

Delivery information: Boxes come with inserts and hinged lids.

 
  • Type:
    Box with insert and lid; holds 4 of 12.5mmx12.5mm mounts
  • Description:
    Mount Holder Box for JEOL 840
  • Size:
    10/pk

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.