Grid Storage Box, 50 Capacity, Electron Microscopy Sciences
Product Details & Documents
Storage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
Documents
EMS-Core Sampling Tools
Product Literature
Specifications
- Capacity:50 Grids
- Description:Grid Storage Box, Clear Cover
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)