Orders LinkContactUsLinkComponent
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High precision diamond scriber, bent tip, 0.01" tip, 0.006" tip length
  100497-826
 :  62105-B2
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
  100497-826
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High precision diamond scriber, bent tip, 0.01" tip, 0.006" tip length
$257.47
Each
A newly designed diamond scribing tool set containing eight precision natural diamond scribes in various sizes and configurations. This set is perfect for precision work in microscopy. The set consists of eight diamond tips, 3 mm in diameter and are mounted in interchangeable tool cones and an MT-1 tool handle. The tool and handle are all housed in a hardwood instrument case. Individual scribers can be purchased separately.
  • Description:
    High precision diamond scriber, bent tip
  • Length:
    0.006" tip length
  • Diameter:
    0.01" tip
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.