Tissue Processing Capsules, Electron Microscopy Sciences
Product Details & Documents
These one piece plastic capsules are hinged and perfectly aligned, offering convenience and ease of use
Both the top and the bottom of the capsule have an open mesh offering maximum fluid flow during processing. A frosted marking surface assures permanent identification. These capsules are commonly used with the Embedding Ring System. They are available in two sizes and five colors.
Specifications
- Depth:8 mm
- Ø:38 mm
- Color:White
- Description:Tissue Processing Capsule for Normal Tissue
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



