Orders LinkContactUsLinkComponent
Cryogenic Vial Rack, Electron Microscopy Sciences
Cryogenic Vial Rack, Electron Microscopy Sciences
Cryogenic Vial Rack
Catalog # 100490-108
Supplier #:  62015-04
Cryogenic Vial Rack, Electron Microscopy Sciences
Catalog # 100490-108
Cryogenic Vial Rack, Electron Microscopy Sciences
Cryogenic Vial Rack, Electron Microscopy Sciences
Cryogenic Vial Rack
Price
$294.96
Log in for availability
Each Pack of 4
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from durable polycarbonate. Its unique design with slotted cavities accepts vials with slotted skirts. When placed in the holes the vials become locked in position, enabling the user to attach or remove the cap with one hand. It accommodates up to 50 vials(1/2" or 13.5 mm diameter) in a 4x10 array with alphanumeric indexing. Autoclavable at 121°C. Measures: 190 x 108 x 22 mm (7 1/2 x 4 1/2 x 7/8").
Specifications
  • Description:
    Cryogenic Vial Rack
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)