Orders LinkContactUsLinkComponent
Safety Bulb, Electron Microscopy Sciences
Safety Bulb, Electron Microscopy Sciences
Safety Bulb
Catalog # 100489-832
Supplier #:  70984
Safety Bulb, Electron Microscopy Sciences
Catalog # 100489-832
Safety Bulb, Electron Microscopy Sciences
Safety Bulb, Electron Microscopy Sciences
Safety Bulb
Price
$173.91
Log in for availability
Each Pack of 4
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Natural rubber bulb allows safe, manual control of filling or discharging toxic liquids through a pipette by finger pressure, thus avoiding mouth contact.
  • Guaranteed to be consistently flexible
Specifications
  • Description:
    Safety Bulb
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)