Orders LinkContactUsLinkComponent
Glass Microbeakers, Electron Microscopy Sciences
Glass Microbeakers, Electron Microscopy Sciences
7.5 ml, 25 mm, 25 mm
Catalog # 100503-380
Supplier #:  60986
Glass Microbeakers, Electron Microscopy Sciences
Catalog # 100503-380
Glass Microbeakers, Electron Microscopy Sciences
Glass Microbeakers, Electron Microscopy Sciences
7.5 ml, 25 mm, 25 mm
Price
$1,198.74
Log in for availability
Box of 100 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Used for the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Specifications
  • Ext.Ø:
    25 mm
  • Capacity:
    7.5 ml
  • Height:
    25 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)