Grid Mats, Electron Microscopy Sciences
Product Details & Documents
White silicone rubber mats, with numbered compartments.
- Good for organizing grids
- They will not slide or jump between compartments
- Ideal for staining grids
Easy to pick up grids without damaging forcep tips. Mats are available for square and round petri dishes, (100 mm diameter).
Specifications
- Description:Round grid mats
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)