Order Entry
Canada
Orders LinkContactUsLinkComponent
Disposable Base Mold, Electron Microscopy Sciences

Disposable Base Mold, Electron Microscopy Sciences

Disposable Base Mold, 5 mm, 24 x 24 mm
Catalog # 100501-998
Minority or Woman-Owned Business Enterprise
Supplier #:  62352-24
Disposable Base Mold, Electron Microscopy Sciences
Catalog # 100501-998
Minority or Woman-Owned Business Enterprise
Disposable Base Mold, Electron Microscopy Sciences
Disposable Base Mold, Electron Microscopy Sciences

Disposable Base Mold, 5 mm, 24 x 24 mm

Price
$280.08
Log in for availability
Each Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Our Disposable Base Mold is designed to fit all existing cassettes. Because our base molds are disposable you save time on cleaning and storing and they offer you less exposure to hazardous chemicals.

Specifications
  • Description:
    Disposable Base Mold
  • Height:
    5 mm
  • Size:
    24 x 24 mm
  • Cat. No.:
    100501-998

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.