High Precision Diamond Scribing Tools, Electron Microscopy Sciences
Product Details & Documents
A newly designed diamond scribing tool set containing eight precision natural diamond scribes in various sizes and configurations. This set is perfect for precision work in microscopy. The set consists of eight diamond tips, 3 mm in diameter and are mounted in interchangeable tool cones and an MT-1 tool handle. The tool and handle are all housed in a hardwood instrument case. Individual scribers can be purchased separately.
Specifications
- Description:High precision diamond scriber
- Length:0.2" tip length
- Diameter:0.02" tip
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)