Precision Screwdriver Set, Electron Microscopy Sciences
Product Details & Documents
The hexagonal swivel heads prevent rolling and knurled handles for a firm grip. Color identifier bands allow for easy selection and set screws hold the blades securely. Set contains 19 extra blades and 2 extra set screws.
Delivery information: The set comes with a handsome revolving stand.
Specifications
- Description:Revolving Stand Screwdriver Set
- Size:—
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.