CoverWell™ Imaging Chambers, Electron Microscopy Sciences
Product Details & Documents
CoverWell™ Imaging chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Specifications
- Depth:0.26 mm
- Description:1 well, 200 µl
- Size:28 mm dia
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)