Orders LinkContactUsLinkComponent
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
8 well, 35 µL, 0.6 mm
Catalog # 100490-788
Supplier #:  70326-50
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
Catalog # 100490-788
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
8 well, 35 µL, 0.6 mm
Price
$379.18
Log in for availability
Each Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CoverWell™ Perfusion Chambers allows up to 16 different specimens on one coverglass.
  • Chambers are removable for permanent mounting of specimens
  • CoverWell™ chambers are made from medical grade silicone bonded to clear, UV transparent plastic
  • Available in multiple designs allowing up to sixteen different specimens or analytical test on one coverglass
  • Ideal for cell imaging on upright or inverted microscopes
"Press-to-Seal" covers form water-tight, multiwall cell incubation or cytochemistry chamber when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports (1.5 mm diameter) without disturbing or cross-contaminating specimens in adjacent wells.
Specifications
  • Depth:
    0.6 mm
  • Description:
    8 well, 35 µL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)