Orders LinkContactUsLinkComponent
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicator, Single End, Non sterile, 100/Pack
Catalog # 100489-878
Supplier #:  72310
Cotton Tipped Applicators, Electron Microscopy Sciences
Catalog # 100489-878
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicator, Single End, Non sterile, 100/Pack
Price
$6.14
Log in for availability
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Lint-free, cotton-tipped applicators are suitable for cleaning delicate parts. Measures 6" in length, 1/12" Ø wooden shaft and head size is 3/16" (Ø) x 5/8" (L).
Handy Vial Packs: 3" (8 cm) wooden shaft, small-tipped applicators come pre-packed in a plastic-capped vial, a handy way to keep a supply of swabs available anytime on the bench top.
Specifications
  • Sterility:
    Non sterile
  • Description:
    Cotton Tipped Applicator, Single End
  • Packaging:
    100/Pack
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.