Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Product Details & Documents
Made from translucent polypropylene.
- Resistant to breakage
- Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
Specifications
- Capacity:5 ml
- Description:Volumetric
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



