Orders LinkContactUsLinkComponent
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Permanox slides
Catalog # 100488-454
Supplier #:  71885
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Catalog # 100488-454
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Permanox™ and Polystyrene Microscope Slides, Electron Microscopy Sciences
Permanox slides
Price
$293.09
Log in for availability
Each Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Note: Long exposure to solvents will cause the slide to warp
Permanox™ is a strong, biologically inert material resistant to non-chlorinated hydrocarbons, acetone, and other EM reagents. Permanox™slides allow you to do cell fixation and staining on the slide; they can be cut with scissors. Permanox™ has not been shown to be autofluorescent. All of the microscope slides are 1 x 3".
Note: Long exposure to solvents will cause the slide to warp. Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.
Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.
Specifications
  • Description:
    Permanox slides
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)