Orders LinkContactUsLinkComponent
Disposable Pipettes – Sterile, Electron Microscopy Sciences
Disposable Pipettes – Sterile, Electron Microscopy Sciences
10 ml, Bulk
Catalog # 101413-250
Supplier #:  70954-10
Disposable Pipettes – Sterile, Electron Microscopy Sciences
Catalog # 101413-250
Disposable Pipettes – Sterile, Electron Microscopy Sciences
Disposable Pipettes – Sterile, Electron Microscopy Sciences
10 ml, Bulk
Price
$68.43
Log in for availability
Each Pack of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to ±1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Specifications
  • Volume:
    10 ml
  • Packed:
    Bulk
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)