StainTray - Slide Staining System, Electron Microscopy Sciences
Product Details & Documents
An ideal device for immunohistochemistry slide staining. The StainTray™ has a black base, made from tough ABS plastic, withstanding a wide range of chemicals (except chlorinated hydrocarbons). It will accept up to 20 slides on four plastic rails covered with a polymer strip to perfectly hold the slides even if the tray is held at an angle.
- Suitable for routine staining requiring a humid chamber
- Ideal for hematology, cytology and microscopy laboratories
- 20-Slide unit with cover: 15 x 93/8 x 13/4" H (38 x 24 x 4.5 cm H)
- 10-Slide unit with cover: 93/8 x 93/8 x 1" (24 x 24 x 4.5 cm H)
When humidity is needed, the wells between the rails will hold the water securely without splashing. Rails are raised not only to avoid water touching the slides but also make the slides more easily retrievable. The base will also hold excess stain solution that drips from the slides. Four rubber feet add to the base for stability. Units are stackable to save space.
A clear cover is available that allows for visual examination. It is made from PETG with temperature a range of -20 to 60 °C. A black cover is available for fluorescent work. It is made from ABS with a temperature range of -80 to 80 °C.
Specifications
- Description:Black Lid Only for 20 Slide Staining Tray
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.