Order Entry
Canada
Orders LinkContactUsLinkComponent
Sodium Cacodylate Buffer, Electron Microscopy Sciences

Sodium Cacodylate Buffer, Electron Microscopy Sciences

Sodium Cacodylate Buffer, 0.2M, Specify, 500 ml
Catalog # 100504-840
Supplier #:  11652
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Catalog # 100504-840
CAS Number:  
Sodium Cacodylate Buffer, Electron Microscopy Sciences
Sodium Cacodylate Buffer, Electron Microscopy Sciences

Sodium Cacodylate Buffer, 0.2M, Specify, 500 ml

CAS Number:  
Price
$146.54
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Used for preparing buffered glutaraldehdye fixatives for electron microscopy

Available with a pH of 7.2 or 7.4.

Sodium Cacodylate Buffer, 0.2M is prepared from Sodium Cacodylate Trihydrate (F.W 214.02).

Documents

Specifications

Specifications

  • Description:
    Sodium Cacodylate Buffer, 0.2M
  • Size:
    500 ml
  • pH:
    Specify

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.