Orders LinkContactUsLinkComponent
Tri-Pour Beaker Caps, Electron Microscopy Sciences
Tri-Pour Beaker Caps, Electron Microscopy Sciences
100 ml (3.4 oz.) beaker
Catalog # 100503-280
Supplier #:  60971
Tri-Pour Beaker Caps, Electron Microscopy Sciences
Catalog # 100503-280
Tri-Pour Beaker Caps, Electron Microscopy Sciences
Tri-Pour Beaker Caps, Electron Microscopy Sciences
100 ml (3.4 oz.) beaker
Price
$138.44
Log in for availability
Each Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
The caps are available for all Tri-Pour beakers.
Specifications
  • Fits:
    100 ml (3.4 oz.) beaker
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.