Orders LinkContactUsLinkComponent
Needle Probe Set, Electron Microscopy Sciences
Replacement Probe #3, 120 mm
  100503-064
 :  62130-03
Needle Probe Set, Electron Microscopy Sciences
  100503-064
Needle Probe Set, Electron Microscopy Sciences
Needle Probe Set, Electron Microscopy Sciences
Replacement Probe #3, 120 mm
$15.51
Each
Set consists of one lightweight precision handle and four interchangeable tools. Can be used for manipulating micro samples, scribing, spring assembly, and probing. The handle is finely balanced, non-rolling gold anodized with split collet and a knurled lock nut. The length is 120 mm (4.7"). Precision forged and ground tools are hardened and nickel plated tool steel with sharp tips.
  • Description:
    Replacement Probe #3
  • Size:
    120 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.