Orders LinkContactUsLinkComponent
Light Pipe Scintillators, Electron Microscopy Sciences
Light Pipe Scintillators, Electron Microscopy Sciences
Cambridge Light Pipe Scintillator, Quartz, Straight, 104 mm
Catalog # 100502-320
Supplier #:  82000-104
Light Pipe Scintillators, Electron Microscopy Sciences
Catalog # 100502-320
Light Pipe Scintillators, Electron Microscopy Sciences
Light Pipe Scintillators, Electron Microscopy Sciences
Cambridge Light Pipe Scintillator, Quartz, Straight, 104 mm
Price
$1,650.57
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT.
  • Scintillator material coated directly on the light pipe which improves the signal
  • Re-coating is typically needed twice per year
Light Pipes are available in optical quality quartz and optical grade acrylic. Cambridge reduced-tip models are only available in acrylic.
Delivery information: Includes one Re-coating Certificate. High voltage wire and corona ring are included, JEOL pipes include a flange.
Specifications
  • Description:
    Cambridge Light Pipe Scintillator, Quartz, Straight, 104 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.