Orders LinkContactUsLinkComponent
Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
100 Capacity, White
Catalog # 100501-436
Supplier #:  71470-W
Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
Catalog # 100501-436
Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
100 Capacity, White
Price
$24.83
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from durable polystyrene (with heavy walls).
This material is unaffected by humidity and is thoroughly insect proof. Each box is secured with a rust-resistant nickel-plated clasp and hinge pin. The cork lining prevents damage to the stored slides. To ensure proper slide identification, each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. To save space the boxes are stackable. Each box stores up to 100 3x1" or 25 x 75 mm microscope slides. available in 5 color coded boxes making the boxes easy to identify. Each box measures 222 x 172 x 35 mm (8.75 x 6.75 x 3.375").
Documents
Specifications
  • Color:
    White
  • Description:
    100 Capacity
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)