Media/Lab Bottles, Electron Microscopy Sciences
Product Details & Documents
Graduated bottles have a large opening, no-drip pour lip and sloping shoulders to facilitate pouring and cleaning.
These bottles are widely used for mixing and storage of culture media, fixation and dehydrating materials. Bottles show approximate volume graduations, and have large frosted writing area. Clear bottles are Wheaton '400' borosilicate glass. Low alkali content helps to prevent changes in pH and maintain purity of contents. Comes with 'PTFE'-lined cap. Autoclavable. Amber bottles are manufactured from '500' borosilicate glass to protect light-sensitive samples.
Specifications
- Capacity:125 ml (4.2 oz.)
- Color:Amber
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.