Orders LinkContactUsLinkComponent
Cryo Markers, Electron Microscopy Sciences
Cryo Markers, Electron Microscopy Sciences
Cryo markers
Catalog # 100500-216
Supplier #:  62051
Cryo Markers, Electron Microscopy Sciences
Catalog # 100500-216
Cryo Markers, Electron Microscopy Sciences
Cryo Markers, Electron Microscopy Sciences
Cryo markers
Price
$248.24
Log in for availability
Case of 24 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Used for permanently marking cloth, cardboard, and plastic.
It will not fade at ultra-low temperatures. Smudge-proof. Package consist of four, extra fine point pens in assorted colors (red, green, blue, and black).
Specifications
  • Description:
    Cryo markers
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)