Orders LinkContactUsLinkComponent
Biopsy Processing Embedding Cassettes, Electron Microscopy Sciences
Biopsy Processing Embedding Cassettes, Electron Microscopy Sciences
Square Mesh Cassettes with Lids Attached, Peach, —
Catalog # 100500-068
Supplier #:  70073-PE
Biopsy Processing Embedding Cassettes, Electron Microscopy Sciences
Catalog # 100500-068
Supplier:  
Biopsy Processing Embedding Cassettes, Electron Microscopy Sciences
Biopsy Processing Embedding Cassettes, Electron Microscopy Sciences
Square Mesh Cassettes with Lids Attached, Peach, —
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CASSETTE BIOPSY W/ LID LAVENDER CS1000 Catalog #76337-612
Product 76337-612 is similar to 100500-068 but may have a different manufacturer and/or packaging.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Biopsy processing cassettes are similar to the slotted cassettes but have a 1 mm square mesh for small biopsies. Made from Acetyl Polymer, which is a special high density polymer that keeps specimens safely submerged in solution, these cassettes are totally resistant to most histological solvents and their slots offer maximum flow through for good fluid exchange and proper drainage.
  • Lids either come attached to the cassette or are packaged separately
  • Lids can be opened and closed as often as necessary and always relock securely without specimen loss
  • Large labeling areas are located on three sides of the cassette for your convenience
  • Each slot measures: 1 x 5 mm, total 128 slots per cassette
Packaging: Cassettes with lids are placed on cassettes, each case contains 3 dispenser boxes of 500 cassettes with lids already locked in place. Cassettes with lids packed separately, each case contains two dispenser boxes of 500 cassettes and Lids.
Specifications
  • Color:
    Peach
  • Description:
    Square Mesh Cassettes with Lids Attached
  • Units:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)