Orders LinkContactUsLinkComponent
Dissecting Needles, Electron Microscopy Sciences
Dissecting Needles, Electron Microscopy Sciences
Curved Dissecting Needle
Catalog # 100499-714
Supplier #:  72949
Dissecting Needles, Electron Microscopy Sciences
Catalog # 100499-714
Dissecting Needles, Electron Microscopy Sciences
Dissecting Needles, Electron Microscopy Sciences
Curved Dissecting Needle
Price
$23.82
Log in for availability
Dozen Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The stainless steel with spear shaped head measures 127 mm (5") long. The wooden handle has a needle tip.
Specifications
  • Description:
    Curved Dissecting Needle
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)