Orders LinkContactUsLinkComponent
Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Ruled Hemacytometer, Complete Set
Catalog # 100498-468
Supplier #:  63511-10
Etched Double Neubauer, Electron Microscopy Sciences
Catalog # 100498-468
Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Ruled Hemacytometer, Complete Set
Price
$1,821.36
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Double improved Neubauer rulings are diamond etched directly into a polished recess to reduce damage from scratches. Opaque surfaces above and below the counting area offer an easy focal reference. The chamber comes with a quick orientation mark for the microscope. 0.1 mm cell depth.
Ordering information: The complete set includes one counting chamber, two 0.5 mm cover glasses, pipettes, and carrying case.
Specifications
  • Description:
    Ruled Hemacytometer, Complete Set
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)