Embedding Blocks, Electron Microscopy Sciences
Product Details & Documents
Embedding blocks are precisely machined from hard phenolic resin
It is suitable for cellodin or paraffin sections. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen.
Specifications
- Description:Embedding Block 4
- Dimensions:51 x 51 x 19 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)