Orders LinkContactUsLinkComponent
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Micro-Needle, 0.25 mm, Tip 1
Catalog # 100497-630
Supplier #:  62091-01-25
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Catalog # 100497-630
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Micro-Needle, 0.25 mm, Tip 1
Price
$150.91
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation.
  • Consists of a threaded handle (MT1), 4.75" long and an interchangeable threaded base with tool tip
  • Available in 32 different tip configurations with interchangeable handles
  • Tip material is hardened tool steel unless noted
Specifications
  • Description:
    Micro-Needle
  • Size:
    0.25 mm
  • Tip configuration:
    Tip 1
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)