Orders LinkContactUsLinkComponent
Hex Key Wrenches, Electron Microscopy Sciences
Hex Key Wrenches, Electron Microscopy Sciences
Hex Key, Metric Set 1
Catalog # 100497-568
Minority or Woman-Owned Business Enterprise
Supplier #:  62076-01
Hex Key Wrenches, Electron Microscopy Sciences
Catalog # 100497-568
Minority or Woman-Owned Business Enterprise
Hex Key Wrenches, Electron Microscopy Sciences
Hex Key Wrenches, Electron Microscopy Sciences
Hex Key, Metric Set 1
Price
$48.21
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from heat treated chrome nickel steel. Useful to have around the lab. Its sturdy handle, folds up just like a pocket knife. Can be used at any angle.
Specifications
  • Description:
    Hex Key, Metric Set 1
  • Cat. No.:
    100497-568
  • Type:
    Hex Key
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)