Flat Mounting Blocks , Electron Microscopy Sciences
Product Details & Documents
Stainless steel flat mounting blocks with the diameter of 25 mm (1") are used for lapping and polishing fixtures.
It is used for the models 150, 151 and 155.
Specifications
- Ø:2"
- Description:Flat Mounting Blocks (Stainless Steel)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)