Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Product Details & Documents
This pen helps to flatten and separate tissue sections and reduces compression in thin sections.
- Powered by AA batteries
Delivery information: Available in two models: Wax Pen 1 is powered by one AA battery; Wax Pen 2 is powered by two AA batteries.
Specifications
- Description:Wax Pen Replacement Tips
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)