Orders LinkContactUsLinkComponent
Count Up/Count Down Timer, Electron Microscopy Sciences
Count Up/Count Down Timer, Electron Microscopy Sciences
Count up/count down timer certified
Catalog # 100493-820
Supplier #:  72603-02
Count Up/Count Down Timer, Electron Microscopy Sciences
Catalog # 100493-820
Count Up/Count Down Timer, Electron Microscopy Sciences
Count Up/Count Down Timer, Electron Microscopy Sciences
Count up/count down timer certified
Price
$173.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Count up or down for 100 hours in intervals of 1 second.
  • Alarm clock mode
  • Its memory allows for the repeat of a cycle just completed
In the count down mode its unique feature tells how much time has elapsed after the alarm sounds.
Dimensions: 2³/₄×2³/₄×¹/₂"
Weight: 1.8 oz.
Certifications: With N.I.S.T. traceable certificate of compliance.
Delivery information: Comes complete with a built in stand, pocket clip, button cell battery, and magnet.
Specifications
  • Description:
    Count up/count down timer certified
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)