Order Entry
Canada
Orders LinkContactUsLinkComponent
Staining Dish Set for 50 Slides, Electron Microscopy Sciences

Staining Dish Set for 50 Slides, Electron Microscopy Sciences

Unit Staining Dish Set
Catalog # 100492-946
Supplier #:  70312-50
Staining Dish Set for 50 Slides, Electron Microscopy Sciences
Catalog # 100492-946
Staining Dish Set for 50 Slides, Electron Microscopy Sciences
Staining Dish Set for 50 Slides, Electron Microscopy Sciences

Unit Staining Dish Set

Price
$762.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The stainless steel, slotted slide rack is designed to hold 50 slides. The rack is made from non-tarnishable stainless steel and is resistant to most staining solutions. The handle is permanently attached allowing for closure of the dish and for easy insertion and removal of the slides.

  • Holds a variety of sizes: 3 x 1", 3 x 11/2" and 3 x 2"
  • Outside: 200 (L) x 105 (W) x 83 (H) mm
  • Inside: 186 (L) x 89 (W) x 68 (H) mm

Delivery information: The complete set consists of one dish, one rack, and one cover.

Specifications
  • Description:
    Unit Staining Dish Set

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.