Orders LinkContactUsLinkComponent
Tissue Chopper, Electron Microscopy Sciences
Tissue Chopper, Electron Microscopy Sciences
TC-1 Tissue Chopper with Micrometer
Catalog # 100492-506
Supplier #:  93102
Tissue Chopper, Electron Microscopy Sciences
Catalog # 100492-506
Tissue Chopper, Electron Microscopy Sciences
Tissue Chopper, Electron Microscopy Sciences
TC-1 Tissue Chopper with Micrometer
Price
$9,250.96
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The TC-1 Tissue Chopper is designed to facilitate the cutting of 100 to 1000 micron thick brain tissue slices for use in recording experiments.
The chopper includes a guillotine arm with adjustments to assure blade alignment. The brain sample is moved in respect to the arm with a stage whose position can be read with 2 to 3 micron precision on a digital micrometre scale. The stage is made with a removable "PTFE" surface plate to facilitate cleaning.
Specifications
  • Description:
    TC-1 Tissue Chopper with Micrometer
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)