Orders LinkContactUsLinkComponent
Developing Trays, Electron Microscopy Sciences
Developing Tray, 20 x 24"
Catalog # 100492-166
 :  74363
Developing Trays, Electron Microscopy Sciences
Catalog # 100492-166
Developing Trays, Electron Microscopy Sciences
Developing Trays, Electron Microscopy Sciences
Developing Tray, 20 x 24"
Price
$372.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These developing trays are made of laminated styrene.
It has wide cross bars and perimeter ribbing that provides rigidity and easy cleaning.
Specifications
  • Description:
    Developing Tray
  • Size:
    20 x 24"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.