Orders LinkContactUsLinkComponent
Scintillation Vial Rack, Electron Microscopy Sciences
Scintillation Vial Rack, Electron Microscopy Sciences
100, 14½×15×2½", Scintilation vial rack, welded
Catalog # 100491-984
Supplier #:  72639
Scintillation Vial Rack, Electron Microscopy Sciences
Catalog # 100491-984
Scintillation Vial Rack, Electron Microscopy Sciences
Scintillation Vial Rack, Electron Microscopy Sciences
100, 14½×15×2½", Scintilation vial rack, welded
Price
$560.78
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Welded polypropylene rack holds 20 ml scintillation vials.
  • Autoclavable
  • Resists nitric and hydrochloric acids
The hole measures ¹¹/₁₆" diameter.
Specifications
  • Description:
    Scintilation vial rack, welded
  • No. of places:
    100
  • W×D×H:
    14½×15×2½"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)