Orders LinkContactUsLinkComponent
Measuring Scale for Measuring Magnifiers, Electron Microscopy Sciences
Measuring Scale for Measuring Magnifiers, Electron Microscopy Sciences
Scale #12, for 10× magnifiers, 35 mm
Catalog # 100491-524
Supplier #:  68184
Measuring Scale for Measuring Magnifiers, Electron Microscopy Sciences
Catalog # 100491-524
Measuring Scale for Measuring Magnifiers, Electron Microscopy Sciences
Measuring Scale for Measuring Magnifiers, Electron Microscopy Sciences
Scale #12, for 10× magnifiers, 35 mm
Price
$45.50
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These scales are available in two different sizes, for use with the measuring magnifiers.
Available in 26 mm for 7× and some for 15× magnifiers and 35 mm for 10× magnifiers.
Specifications
  • Description:
    Scale #12, for 10× magnifiers
  • Size:
    35 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)