Order Entry
Canada
Orders LinkContactUsLinkComponent
Histology Containers, Electron Microscopy Sciences

Histology Containers, Electron Microscopy Sciences

90 ml, Semi-Clear
  100490-446
Supplier #:  64233-16
Histology Containers, Electron Microscopy Sciences
  100490-446
Histology Containers, Electron Microscopy Sciences
Histology Containers, Electron Microscopy Sciences

90 ml, Semi-Clear

 
$484.15
   
Each Pack of 400
 
   
 

The 20 m to 1 L sizes are supplied with tight-fitting screw caps and warning labels for 10% Formalin with extra space for writing

  • Designed for storage and transport of histology specimens
  • Made from durable polypropylene (except 8L size which is made from polyethylene).
  • Leak proof seal caps.

The 2 to 8L sizes are supplied with snap-top lids and carrying handles, and has no graduations.

All of the containers (except the 5 mL size) are graduated on three sides.

 
  • Capacity:
    90 ml
  • Color:
    Semi-Clear

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.