Orders LinkContactUsLinkComponent
Emery Paper, Electron Microscopy Sciences
Emery Paper, Electron Microscopy Sciences
Emery paper, 1/O grit, 400 mesh
Catalog # 100490-128
Supplier #:  62117-01
Emery Paper, Electron Microscopy Sciences
Catalog # 100490-128
Emery Paper, Electron Microscopy Sciences
Emery Paper, Electron Microscopy Sciences
Emery paper, 1/O grit, 400 mesh
Price
$142.71
Log in for availability
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This emery paper comes in sheets and in varying grit sizes.
Grit 4/0 is the finest and 3 is the coarsest.
Size: 22×34 cm (9×13¾")
Specifications
  • Description:
    Emery paper, 1/O grit, 400 mesh
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)