Orders LinkContactUsLinkComponent
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
16 ml, With Plastic Snap Cap, 16 ml
Catalog # 100489-762
Supplier #:  64256
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Catalog # 100489-762
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
16 ml, With Plastic Snap Cap, 16 ml
Price
$536.99
Log in for availability
Each Pack of 144
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Specifications
  • Capacity:
    16 ml
  • Description:
    With Plastic Snap Cap
  • Size:
    16 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.