Microscope Slide Mailer for 5 Slides, CMS Labcraft, Electron Microscopy Sciences
Product Details & Documents
Holds up to 5 slides 3x1"in size . These mailers are rigid, strong and durable with a convenient flip-top, snap-on lid.
When it stands in its upright position, it can be used as a staining container, with a minimal amount of solution being used. Packaged 25 mailers/pk, 8 packs/cs.
Specifications
- Description:Model 1
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)