Orders LinkContactUsLinkComponent
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi-Temp Indicator Tape, ¾" Wide
Catalog # 100488-166
Supplier #:  77015-07
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Catalog # 100488-166
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi-Temp Indicator Tape, ¾" Wide
Price
211,55 $
Log in for availability
Each Roll of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specialty cloth-textured Hi Temperature Indicator tape shows the word “STERILIZED” in bold, black letters after standard sterilization processing. The tape sticks to almost any surface or shape, holds fast in 550°F dry oven temperatures. Available in widths of 1/2", 3/4", and 1" on 250” rolls.
Specifications
  • Description:
    Hi-Temp Indicator Tape, ¾" Wide
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)