Orders LinkContactUsLinkComponent
Cryogenic Labels, Electron Microscopy Sciences
Cryogenic Labels, Electron Microscopy Sciences
Liquistick labels, 0.5×1.75", 500 labels/roll
Catalog # 101413-740
Supplier #:  77027-11
Cryogenic Labels, Electron Microscopy Sciences
Catalog # 101413-740
Supplier:  
Cryogenic Labels, Electron Microscopy Sciences
Cryogenic Labels, Electron Microscopy Sciences
Liquistick labels, 0.5×1.75", 500 labels/roll
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cryogenic labels are made from top quality materials with wide range of sizes, designed specially for cryogenic use.
DuraStick Labels are suitable to be used under a temperature of −40 °C. These labels are features with smudge resistant surface and are available in rolls for thermal transfer printing or in 8½×11" sheets for laser printing. All accept hand written notations with cryo markers.
CryoStick Labels are suitable to be used for temperatures of −80 °C. Ideal for use with cryotubes, these labels are best used with thermal transfer printers or cryo marker pens. Available in a wide range of sizes on sheets or rolls, they are not recommended for laser printing.
LiquiStick Labels are suitable to be used for temperatures of −195 °C. Labels feature with the synthetic surface that accepts crisp bar codes from thermal transfer printers or handwritten notations cryo markers. Supplied in sheets or rolls, these labels are not recommended for use with laser printers.
Satin Cloth Labels are suitable to be used for temperatures of −60 °C. These textured labels withstand dry ice, deep freezers and liquid gases. Supplied in sheets or rolls. They are easily marked with cryo markers, not for use with automated printers.
Wrap-Around Labels are suitable for freezers and cryogenic use. These unique labels are for sub-zero use, measures ¹⁵/₁₆×2³/₈". They feature a ¹⁵/₁₆×¹⁵/₁₆" area, frosted for thermal printing or marking with cryo markers. The remainder of the label is clear. It wraps around the cryo-tube to provide a protective cover for printed information while leaving a clear space for viewing the tube contents.
Specifications
  • Description:
    Liquistick labels
  • Dimensions:
    0.5×1.75"
  • Packaging:
    500 labels/roll
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.