Order Entry
Canada
Orders LinkContactUsLinkComponent
Graduated Transfer Pipettes, Electron Microscopy Sciences

Graduated Transfer Pipettes, Electron Microscopy Sciences

Graduated Transfer Pipettes, 4.8 ml, 7.1×15.2 cm, 2 mL, 1.8 ml, Bulk
Catalog # 101413-284
Supplier #:  70962-10
Graduated Transfer Pipettes, Electron Microscopy Sciences
Catalog # 101413-284
Graduated Transfer Pipettes, Electron Microscopy Sciences
Graduated Transfer Pipettes, Electron Microscopy Sciences

Graduated Transfer Pipettes, 4.8 ml, 7.1×15.2 cm, 2 mL, 1.8 ml, Bulk

Price
$80.57
Log in for availability
Each Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These disposable graduated transfer pipettes are made up of polyethylene.

  • Low-affinity surface
  • Attached bulb avoids cross-contamination
  • One step biohazard disposable

These pipettes are ideal for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, etc.

Specifications
  • Ø×L:
    7.1×15.2 cm
  • Capacity:
    4.8 ml
  • Description:
    Graduated Transfer Pipettes
  • Packed:
    Bulk
  • Graduations:
    2 mL
  • Bulb draw:
    1.8 ml

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.