4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Product Details & Documents
Biopsy processing cassettes with four square compartments each measuring 13 mm.
- Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
Specifications
- Color:Pink
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)