Micromesh™- Single Compartment Biopsy Processing / Embedding Cassettes with Lid, Electron Microscopy Sciences
Product Details & Documents
Made from acetyl polymer This new version of Micromesh™ offers 1676 square openings (0.38 mm) allowing for a greatly improved fluid exchange without having to use the biopsy pads
Each case contains four dispenser boxes of 250 cassettes with covers.
Large anterior and posterior slots in both cassette and cover ensure that the Micromesh™ biopsy cassettes will sink rapidly (far better than cloth mesh cassettes). A large square compartment measuring 27 mm is perfect even for needle biopsies. The cover does not protrude above the cassette, a great space saving feature allowing more cassettes to be stacked in automatic labeling machines and tissue processors.
Specifications
- Color:Gray
- Description:1-Compartment Micromesh™
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)