Orders LinkContactUsLinkComponent
Platinum Crucibles, Electron Microscopy Sciences
Platinum Crucibles, Electron Microscopy Sciences
Platinum Crucible, 15 ml
Catalog # 101413-090
Supplier #:  66504-5
Platinum Crucibles, Electron Microscopy Sciences
Catalog # 101413-090
Platinum Crucibles, Electron Microscopy Sciences
Platinum Crucibles, Electron Microscopy Sciences
Platinum Crucible, 15 ml
Price
$4,483.60
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Platinum has excellent resistance to chemical attack by acids and fusion mixtures. This is what has made platinum an essential tool in chemical analysis; flux fusions which are widely used on a routine basis for the preparation of samples for x-ray fluorescence etc. All crucibles are made from platinum with an approximate weight in grams and an approximate in millimeter with a tolerance of ±5% . Lids are sold separately and in single units.
Ordering information: EMS strives to maintain accurate pricing. However, due to fluctuations in precious metal prices, pricing on products containing precious metals is not guaranteed. We will contact you if there is a discrepancy.
Specifications
  • Capacity:
    15 ml
  • Description:
    Platinum Crucible
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.