Orders LinkContactUsLinkComponent
Lyophilization Seals, Electron Microscopy Sciences
Lyophilization Seals, Electron Microscopy Sciences
Unlined Aluminum Seals, 7x13
Catalog # 101412-414
Supplier #:  60320-07
Lyophilization Seals, Electron Microscopy Sciences
Catalog # 101412-414
Supplier:  
Lyophilization Seals, Electron Microscopy Sciences
Lyophilization Seals, Electron Microscopy Sciences
Unlined Aluminum Seals, 7x13
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Lyophilization seals come in four styles.
The Center Disc Tear Out style exposes the top of the stopper to the syringe. Natural in color. The Flip-Cap, Center Tear style is easy-to-use. Push upward on plastic button to access stopper or septum. No sharp metal edges. Removal of the Flip Cap Button can be accomplished with one hand, freeing the other for the syringe. The Complete Tear Off style seals can be completely removed and contents poured from the bottle. The Lined Aluminum Seals are ideal for filling or retrieving contents with a syringe and as closures for many instrumentation applications. These seals have a center disc tear-out, seal lined with "PTFE" – Faced natural red rubber.
Documents
Specifications
  • Description:
    Unlined Aluminum Seals, 7x13
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)